missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-6-fluorobenzonitrile, TCI America™

$24.44 - $64.17

Chemical Identifiers

CAS 668-45-1
Molecular Formula C7H3ClFN
Molecular Weight (g/mol) 155.556
MDL Number MFCD00001780
InChI Key XPTAYRHLHAFUOS-UHFFFAOYSA-N
Synonym 2-fluoro-6-chlorobenzonitrile, benzonitrile, 2-chloro-6-fluoro, 2-chloro-6-fluoro-benzonitrile, ncr bg ff, pubchem3423, ksc490q6t, acmc-1b036, timtec-bb sbb042501, 2-chloro-6-fluorobenzenecarbonitrile, 6-chloro-2-fluorobenzenecarbonitrile
PubChem CID 69587
IUPAC Name 2-chloro-6-fluorobenzonitrile
SMILES C1=CC(=C(C(=C1)Cl)C#N)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C100225G View Documents TCI America
C100225G
25g Each for $64.17
 
C10025G View Documents TCI America
C10025G
5g Each for $24.44
 
Specifications

Chemical Identifiers

668-45-1
155.556
XPTAYRHLHAFUOS-UHFFFAOYSA-N
69587
C1=CC(=C(C(=C1)Cl)C#N)F
C7H3ClFN
MFCD00001780
2-fluoro-6-chlorobenzonitrile, benzonitrile, 2-chloro-6-fluoro, 2-chloro-6-fluoro-benzonitrile, ncr bg ff, pubchem3423, ksc490q6t, acmc-1b036, timtec-bb sbb042501, 2-chloro-6-fluorobenzenecarbonitrile, 6-chloro-2-fluorobenzenecarbonitrile
2-chloro-6-fluorobenzonitrile

Specifications

White-Yellow
25g
C7H3ClFN
3439
XPTAYRHLHAFUOS-UHFFFAOYSA-N
2-chloro-6-fluorobenzonitrile
69587
Crystal-Powder at 20°C
2-Chloro-6-fluorobenzonitrile
57°C
668-45-1
MFCD00001780
2-fluoro-6-chlorobenzonitrile, benzonitrile, 2-chloro-6-fluoro, 2-chloro-6-fluoro-benzonitrile, ncr bg ff, pubchem3423, ksc490q6t, acmc-1b036, timtec-bb sbb042501, 2-chloro-6-fluorobenzenecarbonitrile, 6-chloro-2-fluorobenzenecarbonitrile
C1=CC(=C(C(=C1)Cl)C#N)F
155.556
155.56
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No