Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-fluorobenzonitrile, 98%

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-fluorobenzonitrile
CAS 668-45-1
Molecular Formula C7H3ClFN
Formula Weight 155.56
MDL Number MFCD00001780
UN Number UN3439
Beilstein 1940332
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2166209

 
alfa aesar™
B2166209
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2166218

 
alfa aesar™
B2166218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 155.56
Chemical Name or Material 2-Chloro-6-fluorobenzonitrile
Odor Characteristic
Assay 98%
Molecular Formula C7H3ClFN
MDL Number MFCD00001780
CAS 668-45-1
UN Number UN3439
Beilstein 1940332
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Boiling Point 104° to 105°C (11mmHg)
Melting Point 56° to 60°C
Quantity 10g
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.