Please login to your online account to display your discounted pricing

2-chloro-6-fluorobenzoic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-fluorobenzoic acid
CAS 434-75-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7 H4 Cl F O2
Linear Formula ClC6H3(F)CO2H
Formula Weight 174.56
MDL Number MFCD00002417
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC176230050

  SDS
thermo fisher scientific
176230050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 174.56
Quantity 5g
Color White to beige
Melting Point 156° to 160°C
CAS Max % 100.0
Infrared Spectrum Authentic
Molecular Formula C7 H4 Cl F O2
Chemical Name or Material 2-Chloro-6-fluorobenzoic acid
Assay Percent Range 98%
MDL Number MFCD00002417
CAS 434-75-3
CAS Min % 97.5
Packaging Glass bottle
Linear Formula ClC6H3(F)CO2H
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.