Please login to your online account to display your discounted pricing

2-chloro-6-fluorobenzoic acid, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  176230050

 View more versions of this product

Catalog No. AC176230050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 174.56
Physical Form Crystalline Powder
Quantity 5g
Color White to beige
Melting Point 156° to 160°C
CAS Max % 100.0
Infrared Spectrum Authentic
Molecular Formula C7 H4 Cl F O2
CAS 434-75-3
Chemical Name or Material 2-Chloro-6-fluorobenzoic acid
Assay Percent Range 98%
MDL Number MFCD00002417
CAS Min % 97.5
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Linear Formula ClC6H3(F)CO2H

Specifications

Chemical Name or Material 2-Chloro-6-fluorobenzoic acid
CAS 434-75-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7 H4 Cl F O2
Linear Formula ClC6H3(F)CO2H
Formula Weight 174.56
MDL Number MFCD00002417
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents