Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-fluorobenzoic acid, 98+%

$34.96 - $459.57

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-fluorobenzoic acid
CAS 434-75-3
Molecular Formula C7H4ClFO2
Formula Weight 174.56
MDL Number MFCD00002417
Beilstein 973857
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1125006

  SDS
alfa aesar™
A1125006
5g Each for $34.96

AAA1125014

  SDS
alfa aesar™
A1125014
25g Each for $126.57

AAA1125022

  SDS
alfa aesar™
A1125022
100g Each for $459.57
Description & Specifications

Specifications

Formula Weight 174.56
Chemical Name or Material 2-Chloro-6-fluorobenzoic acid
Assay ≥98%
Molecular Formula C7H4ClFO2
MDL Number MFCD00002417
CAS 434-75-3
Beilstein 973857
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 156° to 159°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.