Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-fluorobenzaldehyde, 97%

$123.86 - $123.86

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-fluorobenzaldehyde
CAS 387-45-1
Molecular Formula C7H4ClFO
Formula Weight 158.56
MDL Number MFCD00003306
Beilstein 2245530
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1051322

  SDS
alfa aesar™
A1051322
100g Each for $123.86
Description & Specifications

Specifications

Formula Weight 158.56
Chemical Name or Material 2-Chloro-6-fluorobenzaldehyde
TSCA TSCA
Assay 97%
Molecular Formula C7H4ClFO
MDL Number MFCD00003306
CAS 387-45-1
Beilstein 2245530
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 101°C (213°F)
Boiling Point 91° to 93°C (10mmHg)
Melting Point 35° to 39°C
Quantity 100g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.