Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-fluoroanisole, 97+%

$350.96 - $350.96

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-fluoroanisole
CAS 53145-38-3
Molecular Formula C7H6ClFO
Formula Weight 160.58
MDL Number MFCD01631573
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2429406

 
alfa aesar™
B2429406
5g Each for $350.96
Description & Specifications

Specifications

Formula Weight 160.58
Chemical Name or Material 2-Chloro-6-fluoroanisole
Assay ≥97%
Molecular Formula C7H6ClFO
MDL Number MFCD01631573
CAS 53145-38-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.