Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Chloro-6'-fluoroacetophenone, 97%

$72.93 - $988.43

Chemical Identifiers

Chemical Name or Material 2'-Chloro-6'-fluoroacetophenone
CAS 87327-69-3
Molecular Formula C8H6ClFO
Formula Weight 172.59
MDL Number MFCD01631390
Beilstein 5862530
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2406703

  SDS
alfa aesar™
B2406703
1g Each for $72.93

AAB2406714

  SDS
alfa aesar™
B2406714
25g Each for $988.43

AAB2406706

  SDS
alfa aesar™
B2406706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2'-Chloro-6'-fluoroacetophenone
Quantity 1g
CAS 87327-69-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Assay 97%
Molecular Formula C8H6ClFO
Formula Weight 172.59
MDL Number MFCD01631390
Beilstein 5862530

Applications
2′-Chloro-6′-fluoroacetophenone is used in medicine & organic intermediate.

Solubility
Sparingly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.