Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-fluoro-beta-nitrostyrene, 98%

$23.82 - $23.82

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-fluoro-beta-nitrostyrene
CAS 116272-78-7
Molecular Formula C8H5ClFNO2
Formula Weight 201.57g/mol
MDL Number MFCD00053064
Synonym 1-(2-Chloro-6-fluorophenyl)-2-nitroethylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1281503

 
alfa aesar™
A12815-03
1g Each for $23.82
Description & Specifications

Specifications

Melting Point 62° to 64°C
Quantity 1g
Chemical Name or Material 2-Chloro-6-fluoro-beta-nitrostyrene
CAS 116272-78-7
Assay 98%
Molecular Formula C8H5ClFNO2
Formula Weight 201.57g/mol
MDL Number MFCD00053064
Synonym 1-(2-Chloro-6-fluorophenyl)-2-nitroethylene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.