Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-fluoro-3-methylbenzyl alcohol, 97%

$57.58 - $149.31

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-fluoro-3-methylbenzyl alcohol
CAS 261762-83-8
Molecular Formula C8H8ClFO
Formula Weight 174.6
MDL Number MFCD01631362
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1914803

  SDS
alfa aesar™
L1914803
1g Each for $149.31

AAL19148MD

  SDS
alfa aesar™
L19148MD
250mg Each for $57.58
Description & Specifications

Specifications

Formula Weight 174.6
Chemical Name or Material 2-Chloro-6-fluoro-3-methylbenzyl alcohol
Assay 97%
Molecular Formula C8H8ClFO
CAS 261762-83-8
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 68° to 70°C
MDL Number MFCD01631362
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.