Please login to your online account to display your discounted pricing

2-chloro-6-fluoro-3-methylbenzaldehyde, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC376140010

 View more versions of this product

Catalog No. AC376140010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Solid
Formula Weight 172.59
Quantity 1g
Color Off-white
Melting Point 46° to 50°C
CAS Max % 100.0
Linear Formula F(Cl)C6H2(CH3)CHO
Assay Percent Range 98%
Molecular Formula C8H6ClFO
MDL Number MFCD01631360
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Synonym 2-Chloro-6-fluoro-m-tolualdehyde
Infrared Spectrum Authentic
CAS 104451-99-2
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-Chloro-6-fluoro-3-methylbenzaldehyde

Specifications

Chemical Name or Material 2-Chloro-6-fluoro-3-methylbenzaldehyde
CAS 104451-99-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H6ClFO
Linear Formula F(Cl)C6H2(CH3)CHO
Formula Weight 172.59
MDL Number MFCD01631360
Synonym 2-Chloro-6-fluoro-m-tolualdehyde
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents