Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-fluoro-3-methylbenzaldehyde, 97%

$55.89 - $177.80

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-fluoro-3-methylbenzaldehyde
CAS 104451-99-2
Molecular Formula C8H6ClFO
Formula Weight 172.59
MDL Number MFCD01631360
Synonym 2-Chloro-6-fluoro-m-tolualdehyde
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAB2297903

  SDS
alfa aesar™
B2297903
Each for $55.89

AAB2297906

  SDS
alfa aesar™
B2297906
Each for $177.80
Description & Specifications

Specifications

Formula Weight 172.59
MDL Number MFCD01631360
Synonym 2-Chloro-6-fluoro-m-tolualdehyde
Melting Point 26° to 28°C
Flash Point 110°C (230°F)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Chemical Name or Material 2-Chloro-6-fluoro-3-methylbenzaldehyde
Health Hazard 1 Warning
Assay 97%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C8H6ClFO
CAS 104451-99-2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.