Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-fluoro-3-methoxybenzoic acid, 97%

$94.47 - $518.32

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-fluoro-3-methoxybenzoic acid
CAS 886499-40-7
Molecular Formula C8H6ClFO3
Formula Weight 204.59
MDL Number MFCD04115942
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2627603

  SDS
alfa aesar™
H2627603
1g Each for $94.47

AAH2627609

  SDS
alfa aesar™
H2627609
10g Each for $518.32
Description & Specifications

Specifications

Formula Weight 204.59
Assay 97%
Quantity 1g
MDL Number MFCD04115942
Molecular Formula C8H6ClFO3
Chemical Name or Material 2-Chloro-6-fluoro-3-methoxybenzoic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 886499-40-7
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.