Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-fluoro-3-methoxybenzaldehyde, 97%

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-fluoro-3-methoxybenzaldehyde
Molecular Formula C8H6ClFO2
Formula Weight 188.59
MDL Number MFCD04115941
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2640303

  SDS
alfa aesar™
H2640303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2640309

  SDS
alfa aesar™
H2640309
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 188.59
Chemical Name or Material 2-Chloro-6-fluoro-3-methoxybenzaldehyde
Assay 97%
Molecular Formula C8H6ClFO2
MDL Number MFCD04115941
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 104° to 107°C
Quantity 1g

Applications
2-Chloro-6-fluorobenzaldehyde was used in the synthesis of novel copolymers of methyl 2-cyano-3-dihalophenyl-2-propenoates and styrene.

Solubility
Soluble in water 15 mg/L at 20°C . Also soluble in heptane, toluene, methanol, ethyl acetate, acetone.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.