Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-ethoxypyridine, 98%

$89.80 - $1,214.45

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-ethoxypyridine
CAS 42144-78-5
Molecular Formula C7H8ClNO
Formula Weight 157.6
MDL Number MFCD00023465
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2498409

  SDS
alfa aesar™
B2498409
10g Each for $89.80

AAB2498418

  SDS
alfa aesar™
B2498418
50g Each for $320.95

AAB2498430

  SDS
alfa aesar™
B2498430
250g Each for $1,214.45
Description & Specifications

Specifications

Formula Weight 157.6
Chemical Name or Material 2-Chloro-6-ethoxypyridine
Assay 98%
Molecular Formula C7H8ClNO
MDL Number MFCD00023465
CAS 42144-78-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.