Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-cyanopyridine, 97%

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-cyanopyridine
CAS 33252-29-8
Molecular Formula C6H3ClN2
Formula Weight 138.56
MDL Number MFCD00274527
UN Number UN3439
Synonym 6-Chloropicolinonitrile; 6-Chloropyridine-2-carbonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2774803

  SDS
alfa aesar™
H2774803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2774806

  SDS
alfa aesar™
H2774806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 138.56
Chemical Name or Material 2-Chloro-6-cyanopyridine
Assay 97%
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
MDL Number MFCD00274527
Synonym 6-Chloropicolinonitrile; 6-Chloropyridine-2-carbonitrile
Molecular Formula C6H3ClN2
Health Hazard 1 Danger
CAS 33252-29-8
UN Number UN3439
Melting Point 85° to 88°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.