Please login to your online account to display your discounted pricing

2-chloro-5-(trifluoromethyl)pyridine, 98+%, ACROS Organics™

$31.21 - $31.21

Chemical Identifiers

Chemical Name or Material 2-chloro-5-(trifluoromethyl)pyridine
CAS 52334-81-3
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C6H3ClF3N
Formula Weight 181.51
MDL Number MFCD00042225
Synonym 2, 5-CTF
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC241781000

  SDS
thermo fisher scientific
AC241781000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC241780050

  SDS
thermo fisher scientific
241780050
Glass bottle 5g Each for $31.21

AC241780250

  SDS
thermo fisher scientific
241780250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Formula Weight 181.51
Physical Form Crystalline Low Melting Solid
Quantity 100g
Color White to yellow
Melting Point 29° to 33°C
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material 2-chloro-5-(trifluoromethyl)pyridine
Packaging Glass bottle
Molecular Formula C6H3ClF3N
CAS 52334-81-3
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00042225
Assay Percent Range 98+%
Infrared Spectrum Authentic
Synonym 2, 5-CTF

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.