Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-(trifluoromethyl)phenyl isothiocyanate, 97%

Chemical Identifiers

Chemical Name or Material 2-Chloro-5-(trifluoromethyl)phenyl isothiocyanate
CAS 23165-49-3
Molecular Formula C8H3ClF3NS
Formula Weight 237.63
MDL Number MFCD00041055
UN Number UN2922
Beilstein 2112809
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1301603

 
alfa aesar™
L1301603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1301606

 
alfa aesar™
L1301606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 237.63
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)phenyl isothiocyanate
Odor Pungent
Assay 97%
Molecular Formula C8H3ClF3NS
MDL Number MFCD00041055
CAS 23165-49-3
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
UN Number UN2922
Beilstein 2112809
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
Flash Point >110°C (230°F)
Boiling Point 239°C
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.