missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-(trifluoromethyl)phenyl isothiocyanate, 97%

Manufacturer:  Alfa Aesar™ L1301606

 View more versions of this product

Catalog No. AAL1301606



Specifications

Specifications

2-Chloro-5-(trifluoromethyl)phenyl isothiocyanate
Pungent
5g
C8H3ClF3NS
UN2922
2-chloro-5-trifluoromethyl phenyl isothiocyanate, 1-chloro-2-isothiocyanato-4-trifluoromethyl benzene, 2-chloro-5-trifluoromethylphenyl isothiocyanate, 2-chloro-5-trifluoromethyl phenylisothiocyanate, 2-chloro-5-trifluoromethylphenyl isothiocyanate, benzene, 1-chloro-2-isothiocyanato-4-trifluoromethyl, benzene,1-chloro-2-isothiocyanato-4-trifluoromethyl, 2-chloro-5-trifluoromethyl benzenisothiocyanate, pubchem5015, acmc-1ciqp
C1=CC(=C(C=C1C(F)(F)F)N=C=S)Cl
237.624
237.63
>110°C (230°F)
239°C
23165-49-3
MFCD00041055
2112809
KHTMKXDMVYHDSY-UHFFFAOYSA-N
1-chloro-2-isothiocyanato-4-(trifluoromethyl)benzene
141005
97%

Chemical Identifiers

23165-49-3
237.624
KHTMKXDMVYHDSY-UHFFFAOYSA-N
141005
C1=CC(=C(C=C1C(F)(F)F)N=C=S)Cl
C8H3ClF3NS
MFCD00041055
2-chloro-5-trifluoromethyl phenyl isothiocyanate, 1-chloro-2-isothiocyanato-4-trifluoromethyl benzene, 2-chloro-5-trifluoromethylphenyl isothiocyanate, 2-chloro-5-trifluoromethyl phenylisothiocyanate, 2-chloro-5-trifluoromethylphenyl isothiocyanate, benzene, 1-chloro-2-isothiocyanato-4-trifluoromethyl, benzene,1-chloro-2-isothiocyanato-4-trifluoromethyl, 2-chloro-5-trifluoromethyl benzenisothiocyanate, pubchem5015, acmc-1ciqp
1-chloro-2-isothiocyanato-4-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

Documents