Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-(trifluoromethyl)phenol, 98%

Chemical Identifiers

Chemical Name or Material 2-Chloro-5-(trifluoromethyl)phenol
CAS 40889-91-6
Molecular Formula C7H4ClF3O
Formula Weight 196.56
MDL Number MFCD00019995
UN Number UN2924
Beilstein 2094176
Synonym 4-Chloro-3-hydroxybenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0994203

  SDS
alfa aesar™
L0994203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0994206

  SDS
alfa aesar™
L0994206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 196.56
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)phenol
MDL Number MFCD00019995
Assay 98%
CAS 40889-91-6
UN Number UN2924
Synonym 4-Chloro-3-hydroxybenzotrifluoride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H226-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
Molecular Formula C7H4ClF3O
Beilstein 2094176
Flash Point 46°C (114°F)
Boiling Point 87° to 88°C (38mmHg)
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H226-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.