missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-(trifluoromethyl)phenol, 98%

Manufacturer:  Alfa Aesar™ L0994203

 View more versions of this product

Catalog No. AAL0994203



Specifications

Specifications

2-Chloro-5-(trifluoromethyl)phenol
87°C to 88°C (38mmHg)
40889-91-6
MFCD00019995
2094176
SUIZARUATMVYRT-UHFFFAOYSA-N
2-chloro-5-(trifluoromethyl)phenol
123498
98%
46°C (114°F)
1g
C7H4ClF3O
UN2924
2-chloro-5-trifluoromethyl phenol, 4-chloro-3-hydroxybenzotrifluoride, phenol, 2-chloro-5-trifluoromethyl, 2-chloro-5-trifluoromethyl-phenol, 3-hydroxy-4-chlorobenzotrifluoride, m-cresol, 6-chloro-.alpha.,.alpha.,.alpha.-trifluoro-,, pubchem2851, 2-chloro-5-trifluoromethyl phenol, 2-chloranyl-5-trifluoromethyl phenol, 2-chloro-5-trifluoromethyl phenol #
C1=CC(=C(C=C1C(F)(F)F)O)Cl
196.553
196.56

Chemical Identifiers

40889-91-6
196.553
SUIZARUATMVYRT-UHFFFAOYSA-N
123498
C1=CC(=C(C=C1C(F)(F)F)O)Cl
C7H4ClF3O
MFCD00019995
2-chloro-5-trifluoromethyl phenol, 4-chloro-3-hydroxybenzotrifluoride, phenol, 2-chloro-5-trifluoromethyl, 2-chloro-5-trifluoromethyl-phenol, 3-hydroxy-4-chlorobenzotrifluoride, m-cresol, 6-chloro-.alpha.,.alpha.,.alpha.-trifluoro-,, pubchem2851, 2-chloro-5-trifluoromethyl phenol, 2-chloranyl-5-trifluoromethyl phenol, 2-chloro-5-trifluoromethyl phenol #
2-chloro-5-(trifluoromethyl)phenol
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

Documents