missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-(trifluoromethyl)nicotinic acid, 97%

Manufacturer:  Alfa Aesar™ H6335006

 View more versions of this product

Catalog No. AAH6335006



Specifications

Specifications

2-Chloro-5-(trifluoromethyl)nicotinic acid
505084-59-3
C7H3ClF3NO2
2-chloro-5-trifluoromethyl nicotinic acid, 2-chloro-5-trifluoromethyl pyridine-3-carboxylic acid, 2-chloro-5-trifluoromethyl-3-pyridinecarboxylic acid, 2-chloro-5-trifluoromethylnicotinic acid, 2-chloro-5-trifluoromethyl-nicotinic acid, 3-pyridinecarboxylic acid, 2-chloro-5-trifluoromethyl, ksc269c4j, 2-chloro-5-trifluoromethyl nicotinicacid, 2-chloro-5-trifluoromethyl pyridine-3-carboxylic, 2-chloro-5-trifluoromethyl pyridine-3-carboxylicacid
C1=C(C(=NC=C1C(F)(F)F)Cl)C(=O)O
225.551
225.55
97%
173°C to 178°C
5g
MFCD08741353
RMSKYTKOFDQVEL-UHFFFAOYSA-N
2-chloro-5-(trifluoromethyl)pyridine-3-carboxylic acid
12140227
Solid

Chemical Identifiers

505084-59-3
225.551
RMSKYTKOFDQVEL-UHFFFAOYSA-N
12140227
C1=C(C(=NC=C1C(F)(F)F)Cl)C(=O)O
C7H3ClF3NO2
MFCD08741353
2-chloro-5-trifluoromethyl nicotinic acid, 2-chloro-5-trifluoromethyl pyridine-3-carboxylic acid, 2-chloro-5-trifluoromethyl-3-pyridinecarboxylic acid, 2-chloro-5-trifluoromethylnicotinic acid, 2-chloro-5-trifluoromethyl-nicotinic acid, 3-pyridinecarboxylic acid, 2-chloro-5-trifluoromethyl, ksc269c4j, 2-chloro-5-trifluoromethyl nicotinicacid, 2-chloro-5-trifluoromethyl pyridine-3-carboxylic, 2-chloro-5-trifluoromethyl pyridine-3-carboxylicacid
2-chloro-5-(trifluoromethyl)pyridine-3-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents