Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-(trifluoromethyl)benzylamine, 97%

Chemical Identifiers

Chemical Name or Material 2-Chloro-5-(trifluoromethyl)benzylamine
CAS 15996-78-8
Molecular Formula C8H7ClF3N
Formula Weight 209.6
MDL Number MFCD00190123
UN Number UN2735
Beilstein 2835666
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2444903

 
alfa aesar™
B2444903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2444906

 
alfa aesar™
B2444906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 209.6
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)benzylamine
Odor Amine-like
Assay 97%
Molecular Formula C8H7ClF3N
MDL Number MFCD00190123
CAS 15996-78-8
UN Number UN2735
Beilstein 2835666
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Flash Point 100°C (212°F)
Boiling Point 90° to 94°C (13mmHg)
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.