Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-(trifluoromethyl)benzyl alcohol, 97+%

$221.66 - $221.66

Chemical Identifiers

Chemical Name or Material 2-Chloro-5-(trifluoromethyl)benzyl alcohol
CAS 64372-62-9
Molecular Formula C8H6ClF3O
Formula Weight 210.58
MDL Number MFCD01631469
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2393706

 
alfa aesar™
B2393706
5g Each for $221.66
Description & Specifications

Specifications

Formula Weight 210.58
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)benzyl alcohol
Molecular Formula C8H6ClF3O
CAS 64372-62-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay ≥97%
MDL Number MFCD01631469
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 57° to 60°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.