missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-(trifluoromethyl)benzoic acid, 98%

Manufacturer:  Alfa Aesar™ H5544806

 View more versions of this product

Catalog No. AAH5544806



Specifications

Specifications

2-Chloro-5-(trifluoromethyl)benzoic acid
5g
C8H4ClF3O2
2-chloro-5-trifluoromethyl benzoic acid
C1=CC(=C(C=C1C(F)(F)F)C(=O)O)Cl
224.563
224.57
93°C to 96°C
657-06-7
MFCD00061094
WLXRKCGYQAKHSJ-UHFFFAOYSA-N
2-chloro-5-(trifluoromethyl)benzoic acid
688182
98%

Chemical Identifiers

657-06-7
224.563
WLXRKCGYQAKHSJ-UHFFFAOYSA-N
688182
C1=CC(=C(C=C1C(F)(F)F)C(=O)O)Cl
C8H4ClF3O2
MFCD00061094
2-chloro-5-trifluoromethyl benzoic acid
2-chloro-5-(trifluoromethyl)benzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents