Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-(trifluoromethyl)benzaldehyde, 97+%

$51.99 - $153.69

Chemical Identifiers

Chemical Name or Material 2-Chloro-5-(trifluoromethyl)benzaldehyde
CAS 82386-89-8
Molecular Formula C8H4ClF3O
Formula Weight 208.57
MDL Number MFCD00134430
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2292503

  SDS
alfa aesar™
B2292503
1g Each for $51.99

AAB2292506

  SDS
alfa aesar™
B2292506
5g Each for $153.69
Description & Specifications

Specifications

Formula Weight 208.57
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)benzaldehyde
Assay ≥97%
Molecular Formula C8H4ClF3O
MDL Number MFCD00134430
CAS 82386-89-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 89°C (103°F)
Boiling Point 42° to 44°C (1.5mmHg)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.