missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-(trifluoromethoxy)benzeneboronic acid, 98%

$43.03 - $171.18

Chemical Identifiers

CAS 1022922-16-2
Molecular Formula C7H5BClF3O3
Molecular Weight (g/mol) 240.369
MDL Number MFCD06656267
InChI Key BAEHYECKXGVYKR-UHFFFAOYSA-N
Synonym 2-chloro-5-trifluoromethoxy phenylboronic acid, 2-chloro-5-trifluoromethoxy phenyl boronic acid, 2-chloro-5-trifluoromethoxyphenylboronic acid, 2-chloro-5-trifluoromethoxy phenyl boronic acid, boronic acid, b-2-chloro-5-trifluoromethoxy phenyl, pubchem23778, acmc-1c1ll, ksc500s0d, 2-chloro-5-trifluoromethoxy phenylboronicacid, 2-chloro-5-trifluoromethoxy benzeneboronic acid
PubChem CID 2763238
IUPAC Name [2-chloro-5-(trifluoromethoxy)phenyl]boronic acid
SMILES B(C1=C(C=CC(=C1)OC(F)(F)F)Cl)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6434103 View Documents Alfa Aesar™
H6434103
1g Each for $43.03
 
AAH6434106 View Documents Alfa Aesar™
H6434106
5g Each for $171.18
 
Specifications

Chemical Identifiers

1022922-16-2
240.369
BAEHYECKXGVYKR-UHFFFAOYSA-N
2763238
B(C1=C(C=CC(=C1)OC(F)(F)F)Cl)(O)O
C7H5BClF3O3
MFCD06656267
2-chloro-5-trifluoromethoxy phenylboronic acid, 2-chloro-5-trifluoromethoxy phenyl boronic acid, 2-chloro-5-trifluoromethoxyphenylboronic acid, 2-chloro-5-trifluoromethoxy phenyl boronic acid, boronic acid, b-2-chloro-5-trifluoromethoxy phenyl, pubchem23778, acmc-1c1ll, ksc500s0d, 2-chloro-5-trifluoromethoxy phenylboronicacid, 2-chloro-5-trifluoromethoxy benzeneboronic acid
[2-chloro-5-(trifluoromethoxy)phenyl]boronic acid

Specifications

1g
C7H5BClF3O3
2-chloro-5-trifluoromethoxy phenylboronic acid, 2-chloro-5-trifluoromethoxy phenyl boronic acid, 2-chloro-5-trifluoromethoxyphenylboronic acid, 2-chloro-5-trifluoromethoxy phenyl boronic acid, boronic acid, b-2-chloro-5-trifluoromethoxy phenyl, pubchem23778, acmc-1c1ll, ksc500s0d, 2-chloro-5-trifluoromethoxy phenylboronicacid, 2-chloro-5-trifluoromethoxy benzeneboronic acid
B(C1=C(C=CC(=C1)OC(F)(F)F)Cl)(O)O
240.369
240.37
98%
1022922-16-2
MFCD06656267
BAEHYECKXGVYKR-UHFFFAOYSA-N
[2-chloro-5-(trifluoromethoxy)phenyl]boronic acid
2763238
Solid
2-Chloro-5-(trifluoromethoxy)benzeneboronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning