Please login to your online account to display your discounted pricing

2-chloro-5-pyrimidinecarbonyl chloride, 95%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC441310010

 View more versions of this product

Catalog No. AC441310010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94.0%
Formula Weight 176.99
Physical Form Powder or Low Melting Solid
Quantity 1g
Color White to yellow
Melting Point 49° to 54°C
CAS Max % 100.0
CAS Min % 94.0
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.
Chemical Name or Material 2-Chloro-5-pyrimidinecarbonyl chloride
Assay Percent Range 95%
CAS 110099-99-5
Molecular Formula C5H2Cl2N2O
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material 2-Chloro-5-pyrimidinecarbonyl chloride
CAS 110099-99-5
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94.0%
Assay Percent Range 95%
Molecular Formula C5H2Cl2N2O
Formula Weight 176.99
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.

SDS & Safety Documents