missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-chloro-5-pyrimidinecarbonyl chloride, 95%, ACROS Organics™

Chemical Identifiers

CAS 110099-99-5
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C5H2Cl2N2O
Formula Weight 176.99
Chemical Name or Material 2-Chloro-5-pyrimidinecarbonyl chloride
Assay ≥94.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC441310010 SDS Thermo Fisher Scientific
AC441310010
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC441310050 SDS Thermo Fisher Scientific
AC441310050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

110099-99-5
100.0
C5H2Cl2N2O
2-Chloro-5-pyrimidinecarbonyl chloride
94.0
95%
176.99
≥94.0%

Specifications

176.99
1g
49° to 54°C
94.0
110099-99-5
Authentic
≥94.0%
Powder or Low Melting Solid
White to yellow
100.0
95%
C5H2Cl2N2O
2-Chloro-5-pyrimidinecarbonyl chloride

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger