Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-nitrotoluene, 98+%

Chemical Identifiers

Chemical Name or Material 2-Chloro-5-nitrotoluene
CAS 13290-74-9
Molecular Formula C7H6ClNO2
Formula Weight 171.58
MDL Number MFCD00041430
UN Number UN3457
Beilstein 2044766
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1083206

  SDS
alfa aesar™
L1083206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1083214

  SDS
alfa aesar™
L1083214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 171.58
Chemical Name or Material 2-Chloro-5-nitrotoluene
TSCA TSCA
Assay ≥98%
MDL Number MFCD00041430
CAS 13290-74-9
Beilstein 2044766
Health Hazard 2 GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLORONITROTOLUENES, SOLID
Melting Point 42° to 44°C
Molecular Formula C7H6ClNO2
UN Number UN3457
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 94° to 96°C (4mmHg)
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.