missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-nitrotoluene, 98+%

Manufacturer:  Alfa Aesar™ L1083206

 View more versions of this product

Catalog No. AAL1083206



Specifications

Specifications

2-Chloro-5-nitrotoluene
42°C to 44°C
13290-74-9
MFCD00041430
2044766
BGDCQZFFNFXYQC-UHFFFAOYSA-N
1-chloro-2-methyl-4-nitrobenzene
83315
≥98%
94°C to 96°C (4mmHg)
5g
C7H6ClNO2
UN3457
2-chloro-5-nitrotoluene, benzene, 1-chloro-2-methyl-4-nitro, 4-chloro-3-methylnitrobenzene, 1-chloro-2-methyl-4-nitro-benzene, 3-nitro-6-chlorotoluene, 2-methyl-4-nitrochlorobenzene, pubchem2137, 2-chloro-5-nitroluene, acmc-209bqf, 5-nitro-2-chlorotoluene
CC1=C(C=CC(=C1)[N+](=O)[O-])Cl
171.58
171.58

Chemical Identifiers

13290-74-9
171.58
BGDCQZFFNFXYQC-UHFFFAOYSA-N
83315
CC1=C(C=CC(=C1)[N+](=O)[O-])Cl
C7H6ClNO2
MFCD00041430
2-chloro-5-nitrotoluene, benzene, 1-chloro-2-methyl-4-nitro, 4-chloro-3-methylnitrobenzene, 1-chloro-2-methyl-4-nitro-benzene, 3-nitro-6-chlorotoluene, 2-methyl-4-nitrochlorobenzene, pubchem2137, 2-chloro-5-nitroluene, acmc-209bqf, 5-nitro-2-chlorotoluene
1-chloro-2-methyl-4-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLORONITROTOLUENES, SOLID

TSCA : TSCA

Documents