Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-nitrophenyl isocyanate, 96%

Chemical Identifiers

Chemical Name or Material 2-Chloro-5-nitrophenyl isocyanate
CAS 68622-16-2
Molecular Formula C7H3ClN2O3
Formula Weight 198.57
MDL Number MFCD00037065
UN Number UN2811
Beilstein 6506805
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1599403

  SDS
alfa aesar™
L1599403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1599406

  SDS
alfa aesar™
L1599406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 198.57
MDL Number MFCD00037065
CAS 68622-16-2
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
UN Number UN2811
Melting Point 92° to 95°C
Molecular Formula C7H3ClN2O3
Assay 96%
Health Hazard 1 Danger
Beilstein 6506805
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Health Hazard 2 GHS H Statement
H331-H334-H302-H312-H315-H319-H317-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Chemical Name or Material 2-Chloro-5-nitrophenyl isocyanate
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H334-H302-H312-H315-H319-H317-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.