Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-nitronicotinic acid, 95%

Chemical Identifiers

Chemical Name or Material 2-Chloro-5-nitronicotinic acid
CAS 42959-38-6
Formula Weight 202.55
MDL Number MFCD06658402
Synonym 2-Chloro-5-nitropyridine-3-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6692703

  SDS
alfa aesar™
H6692703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6692706

  SDS
alfa aesar™
H6692706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 140° to 143°C
Formula Weight 202.55
Assay 95%
Chemical Name or Material 2-Chloro-5-nitronicotinic acid
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD06658402
Health Hazard 1 Warning
Color White to yellow
Quantity 1g
Physical Form Solid
Synonym 2-Chloro-5-nitropyridine-3-carboxylic acid
Sensitivity Moisture Sensitive
CAS 42959-38-6
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.