missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-nitrocinnamic acid, predominantly trans, 98%

Manufacturer:  Alfa Aesar™ A1472706

 View more versions of this product

Catalog No. AAA1472706



Specifications

Specifications

2-Chloro-5-nitrocinnamic acid
5g
36015-19-7
MFCD00063312
2-chloro-5-nitrocinnamic acid, 2e-3-2-chloro-5-nitrophenyl prop-2-enoic acid, 3-2-chloro-5-nitrophenyl acrylic acid, 3-2-chloro-5-nitrophenyl propenoic acid, 2-chloro-5-nitrocinnamic acid, predominantly trans, 2e-3-2-chloro-5-nitrophenyl-2-propenoic acid, e-3-2-chloro-5-nitrophenyl prop-2-enoic acid, 2e-3-2-chloro-5-nitrophenyl-2-propenoic acid #
C1=CC(=C(C=C1[N+](=O)[O-])C=CC(=O)[O-])Cl
226.592
227.61
220°C to 224°C
predominantly trans
C9H5ClNO4-
1972431
IHTLKLSRRSWIGD-DAFODLJHSA-M
(E)-3-(2-chloro-5-nitrophenyl)prop-2-enoate
6921604
98%

Chemical Identifiers

36015-19-7
226.592
IHTLKLSRRSWIGD-DAFODLJHSA-M
6921604
C1=CC(=C(C=C1[N+](=O)[O-])C=CC(=O)[O-])Cl
C9H5ClNO4-
MFCD00063312
2-chloro-5-nitrocinnamic acid, 2e-3-2-chloro-5-nitrophenyl prop-2-enoic acid, 3-2-chloro-5-nitrophenyl acrylic acid, 3-2-chloro-5-nitrophenyl propenoic acid, 2-chloro-5-nitrocinnamic acid, predominantly trans, 2e-3-2-chloro-5-nitrophenyl-2-propenoic acid, e-3-2-chloro-5-nitrophenyl prop-2-enoic acid, 2e-3-2-chloro-5-nitrophenyl-2-propenoic acid #
(E)-3-(2-chloro-5-nitrophenyl)prop-2-enoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents