Please login to your online account to display your discounted pricing

2-chloro-5-nitrobenzyl alcohol, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-chloro-5-nitrobenzyl alcohol
CAS 80866-80-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H6ClNO3
Linear Formula ClC6H3(NO2)CH2OH
Formula Weight 187.58
MDL Number MFCD00007297
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC182930020

  SDS
thermo fisher scientific
182930020
Glass bottle 2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder
Formula Weight 187.58
Quantity 2g
Color Beige
Melting Point 74° to 79°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2-chloro-5-nitrobenzyl alcohol
Assay Percent Range 97%
Linear Formula ClC6H3(NO2)CH2OH
MDL Number MFCD00007297
CAS 80866-80-4
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C7H6ClNO3
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.