Please login to your online account to display your discounted pricing

2-chloro-5-nitrobenzyl alcohol, 97%, ACROS Organics™


Quantity 2g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  182930020

 View more versions of this product

Catalog No. AC182930020

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Powder
Formula Weight 187.58
Quantity 2g
Color Beige
Melting Point 74° to 79°C
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Linear Formula ClC6H3(NO2)CH2OH
CAS 80866-80-4
Chemical Name or Material 2-chloro-5-nitrobenzyl alcohol
Packaging Glass bottle
Assay Percent Range 97%
MDL Number MFCD00007297
Molecular Formula C7H6ClNO3
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material 2-chloro-5-nitrobenzyl alcohol
CAS 80866-80-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H6ClNO3
Linear Formula ClC6H3(NO2)CH2OH
Formula Weight 187.58
MDL Number MFCD00007297
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents