missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-nitrobenzoyl chloride, 96%

$88.39 - $242.69

Chemical Identifiers

CAS 25784-91-2
Molecular Formula C7H3Cl2NO3
Molecular Weight (g/mol) 220.005
MDL Number MFCD00059180
InChI Key OGLKKYALUKXVPQ-UHFFFAOYSA-N
Synonym 2-chloranyl-5-nitro-benzoyl chloride
PubChem CID 117637
IUPAC Name 2-chloro-5-nitrobenzoyl chloride
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(=O)Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1752314 View Documents Alfa Aesar™
L1752314
25g Each for $88.39
 
AAL1752322 View Documents Alfa Aesar™
L1752322
100g Each for $242.69
 
Specifications

Chemical Identifiers

25784-91-2
220.005
OGLKKYALUKXVPQ-UHFFFAOYSA-N
117637
C1=CC(=C(C=C1[N+](=O)[O-])C(=O)Cl)Cl
C7H3Cl2NO3
MFCD00059180
2-chloranyl-5-nitro-benzoyl chloride
2-chloro-5-nitrobenzoyl chloride

Specifications

Moisture sensitive
58°C to 60°C
25784-91-2
MFCD00059180
2213721
OGLKKYALUKXVPQ-UHFFFAOYSA-N
2-chloro-5-nitrobenzoyl chloride
117637
96%
157°C to 158°C (11mmHg)
25g
C7H3Cl2NO3
UN3261
2-chloranyl-5-nitro-benzoyl chloride
C1=CC(=C(C=C1[N+](=O)[O-])C(=O)Cl)Cl
220.005
220.01
2-Chloro-5-nitrobenzoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 247-262-4

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen