missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-nitrobenzoyl chloride, 96%

Manufacturer:  Alfa Aesar™ L1752322

 View more versions of this product

Catalog No. AAL1752322



Specifications

Specifications

2-Chloro-5-nitrobenzoyl chloride
157°C to 158°C (11mmHg)
100g
C7H3Cl2NO3
UN3261
2-chloranyl-5-nitro-benzoyl chloride, 2-chloro-5-nitro-benzoic chloride, 2-chloro-5-nitro-benzoyl chloride, 2-chloro-5-nitro-benzoylchlorid, 2-chloro-5-nitrobenzoic acid chloride, 2-chloro-5-nitrobenzoylchloride, 2-chloro-5nitrobenzoyl chloride, acmc-1cq6z, benzoyl chloride, 2-chloro-5-nitro, benzoyl chloride,2-chloro-5-nitro
C1=CC(=C(C=C1[N+](=O)[O-])C(=O)Cl)Cl
220.005
220.01
Moisture sensitive
58°C to 60°C
25784-91-2
MFCD00059180
2213721
OGLKKYALUKXVPQ-UHFFFAOYSA-N
2-chloro-5-nitrobenzoyl chloride
117637
96%

Chemical Identifiers

25784-91-2
220.005
OGLKKYALUKXVPQ-UHFFFAOYSA-N
117637
C1=CC(=C(C=C1[N+](=O)[O-])C(=O)Cl)Cl
C7H3Cl2NO3
MFCD00059180
2-chloranyl-5-nitro-benzoyl chloride, 2-chloro-5-nitro-benzoic chloride, 2-chloro-5-nitro-benzoyl chloride, 2-chloro-5-nitro-benzoylchlorid, 2-chloro-5-nitrobenzoic acid chloride, 2-chloro-5-nitrobenzoylchloride, 2-chloro-5nitrobenzoyl chloride, acmc-1cq6z, benzoyl chloride, 2-chloro-5-nitro, benzoyl chloride,2-chloro-5-nitro
2-chloro-5-nitrobenzoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 247-262-4

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents