Please login to your online account to display your discounted pricing

2-Chloro-5-nitrobenzaldehyde, +99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC404870250

 View more versions of this product

Catalog No. AC404870250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥98.5%
Formula Weight 185.57
Quantity 25g
Color Pale yellow
EINECS Number 228-830-0
TSCA TSCA
Melting Point 77° to 81°C
RTECS Number CU5220000
CAS Max % 100.0
Chemical Name or Material 2-Chloro-5-nitrobenzaldehyde
Name Note 99+%
Assay Percent Range 99%
Molecular Formula C7H4ClNO3
Linear Formula ClC6H3(NO2)CHO
MDL Number MFCD00007293
CAS 6361-21-3
CAS Min % 98.5
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

Specifications

Chemical Name or Material 2-Chloro-5-nitrobenzaldehyde
Name Note 99+%
CAS 6361-21-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H4ClNO3
Linear Formula ClC6H3(NO2)CHO
Formula Weight 185.57
MDL Number MFCD00007293
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents