missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-5-nitrobenzaldehyde 97%, Acros Organics

Manufacturer:  Acros Organics 160631000

 View more versions of this product

Catalog No. AC160631000



Specifications

Specifications

2-Chloro-5-nitrobenzaldehyde
Beige-Yellow to Yellow
100g
6361-21-3
100.0
C7H4ClNO3
07, 262
VFVHWCKUHAEDMY-UHFFFAOYSA-N
2-chloro-5-nitrobenzaldehyde
72933
Crystalline Powder
Authentic
76°C to 82°C
97%
96.0
97%
ClC6H3(NO2)CHO
benzaldehyde, 2-chloro-5-nitro, 3-nitro-6-chlorobenzaldehyde, 2-chloro-5-nitro-benzaldehyde, unii-pf4ill0i3h, pf4ill0i3h, 2-chlor-5-nitrobenzaldehyd, pubchem7375, wln: wnr dg cvh, chloro-5-nitrobenzaldehyde, asischem r37046
C1=CC(=C(C=C1[N+](=O)[O-])C=O)Cl
185.563
185.57
≥96.0%

Chemical Identifiers

6361-21-3
185.563
benzaldehyde, 2-chloro-5-nitro, 3-nitro-6-chlorobenzaldehyde, 2-chloro-5-nitro-benzaldehyde, unii-pf4ill0i3h, pf4ill0i3h, 2-chlor-5-nitrobenzaldehyd, pubchem7375, wln: wnr dg cvh, chloro-5-nitrobenzaldehyde, asischem r37046
2-chloro-5-nitrobenzaldehyde
C7H4ClNO3
VFVHWCKUHAEDMY-UHFFFAOYSA-N
72933
C1=CC(=C(C=C1[N+](=O)[O-])C=O)Cl
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 228-830-0

RTECSNumber : CU5220000

TSCA : TSCA

SDS Documents