missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-5-nitrobenzaldehyde 97%, Acros Organics

$431.76 - $431.76

Chemical Identifiers

CAS 6361-21-3
Molecular Formula C7H4ClNO3
Molecular Weight (g/mol) 185.563
InChI Key VFVHWCKUHAEDMY-UHFFFAOYSA-N
Synonym benzaldehyde, 2-chloro-5-nitro, 3-nitro-6-chlorobenzaldehyde, 2-chloro-5-nitro-benzaldehyde, unii-pf4ill0i3h, pf4ill0i3h, 2-chlor-5-nitrobenzaldehyd, pubchem7375, wln: wnr dg cvh, chloro-5-nitrobenzaldehyde, asischem r37046
PubChem CID 72933
IUPAC Name 2-chloro-5-nitrobenzaldehyde
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC160631000 SDS View Documents Acros Organics
160631000
100g Each for $431.76
Specifications

Chemical Identifiers

6361-21-3
185.563
benzaldehyde, 2-chloro-5-nitro, 3-nitro-6-chlorobenzaldehyde, 2-chloro-5-nitro-benzaldehyde, unii-pf4ill0i3h, pf4ill0i3h, 2-chlor-5-nitrobenzaldehyd, pubchem7375, wln: wnr dg cvh, chloro-5-nitrobenzaldehyde, asischem r37046
2-chloro-5-nitrobenzaldehyde
C7H4ClNO3
VFVHWCKUHAEDMY-UHFFFAOYSA-N
72933
C1=CC(=C(C=C1[N+](=O)[O-])C=O)Cl

Specifications

Authentic
76°C to 82°C
6361-21-3
100.0
C7H4ClNO3
07, 262
VFVHWCKUHAEDMY-UHFFFAOYSA-N
2-chloro-5-nitrobenzaldehyde
72933
Crystalline Powder
2-Chloro-5-nitrobenzaldehyde, 97%
Beige-Yellow to Yellow
100g
96.0
97%
ClC6H3(NO2)CHO
benzaldehyde, 2-chloro-5-nitro, 3-nitro-6-chlorobenzaldehyde, 2-chloro-5-nitro-benzaldehyde, unii-pf4ill0i3h, pf4ill0i3h, 2-chlor-5-nitrobenzaldehyd, pubchem7375, wln: wnr dg cvh, chloro-5-nitrobenzaldehyde, asischem r37046
C1=CC(=C(C=C1[N+](=O)[O-])C=O)Cl
185.563
185.57
≥96.0%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 228-830-0

RTECSNumber : CU5220000

TSCA : TSCA