Learn More
2-Chloro-5-nitrobenzaldehyde, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1482418
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Chloro-5-nitrobenzaldehyde | |
77°C to 81°C | |
MFCD00007293 | |
743764 | |
benzaldehyde, 2-chloro-5-nitro, 3-nitro-6-chlorobenzaldehyde, 2-chloro-5-nitro-benzaldehyde, unii-pf4ill0i3h, pf4ill0i3h, 2-chlor-5-nitrobenzaldehyd, pubchem7375, wln: wnr dg cvh, chloro-5-nitrobenzaldehyde, asischem r37046 | |
[O-][N+](=O)C1=CC=C(Cl)C(C=O)=C1 | |
185.56 | |
185.57 |
6361-21-3 | |
C7H4ClNO3 | |
50 g | |
Air Sensitive | |
VFVHWCKUHAEDMY-UHFFFAOYSA-N | |
2-chloro-5-nitrobenzaldehyde | |
72933 | |
97% |
Chemical Identifiers
6361-21-3 | |
185.56 | |
VFVHWCKUHAEDMY-UHFFFAOYSA-N | |
72933 | |
[O-][N+](=O)C1=CC=C(Cl)C(C=O)=C1 |
C7H4ClNO3 | |
MFCD00007293 | |
benzaldehyde, 2-chloro-5-nitro, 3-nitro-6-chlorobenzaldehyde, 2-chloro-5-nitro-benzaldehyde, unii-pf4ill0i3h, pf4ill0i3h, 2-chlor-5-nitrobenzaldehyd, pubchem7375, wln: wnr dg cvh, chloro-5-nitrobenzaldehyde, asischem r37046 | |
2-chloro-5-nitrobenzaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 228-830-0
RTECSNumber : CU5220000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only