missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-nitrobenzaldehyde, 97%

Manufacturer:  Alfa Aesar™ A1482422

 View more versions of this product

Catalog No. AAA1482422



Specifications

Specifications

2-Chloro-5-nitrobenzaldehyde
100g
C7H4ClNO3
743764
VFVHWCKUHAEDMY-UHFFFAOYSA-N
2-chloro-5-nitrobenzaldehyde
72933
97%
77°C to 81°C
6361-21-3
MFCD00007293
benzaldehyde, 2-chloro-5-nitro, 3-nitro-6-chlorobenzaldehyde, 2-chloro-5-nitro-benzaldehyde, unii-pf4ill0i3h, pf4ill0i3h, 2-chlor-5-nitrobenzaldehyd, pubchem7375, wln: wnr dg cvh, chloro-5-nitrobenzaldehyde, asischem r37046
C1=CC(=C(C=C1[N+](=O)[O-])C=O)Cl
185.563
185.57

Chemical Identifiers

6361-21-3
185.563
VFVHWCKUHAEDMY-UHFFFAOYSA-N
72933
C1=CC(=C(C=C1[N+](=O)[O-])C=O)Cl
C7H4ClNO3
MFCD00007293
benzaldehyde, 2-chloro-5-nitro, 3-nitro-6-chlorobenzaldehyde, 2-chloro-5-nitro-benzaldehyde, unii-pf4ill0i3h, pf4ill0i3h, 2-chlor-5-nitrobenzaldehyd, pubchem7375, wln: wnr dg cvh, chloro-5-nitrobenzaldehyde, asischem r37046
2-chloro-5-nitrobenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : CU5220000

TSCA : TSCA

SDS Documents