missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-5-nitroaniline, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 109575000

 View more versions of this product

Catalog No. AC109575000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Chloro-5-nitroaniline
6283-25-6
Authentic
Glass bottle
500g
Powder
99%
ClC6H3(NO2)NH2
MFCD00007668
99%
98.5
100.0
118° to 121°C
Yellow
≥98.5%
C6H5ClN2O2
172.57
12, 732

Chemical Identifiers

6283-25-6
100.0
C6H5ClN2O2
172.57
2-Chloro-5-nitroaniline, 99%
98.5
99%
ClC6H3(NO2)NH2
MFCD00007668
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Fatal in contact with skin.
Fatal if swallowed.
Fatal if inhaled.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 228-498-7

RTECSNumber : BX1500000

TSCA : TSCA