missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-5-nitroaniline, 99%, ACROS Organics™

Chemical Identifiers

CAS 6283-25-6
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.568
MDL Number MFCD00007668
InChI Key KWIXNFOTNVKIGM-UHFFFAOYSA-N
Synonym benzenamine, 2-chloro-5-nitro, 6-chloro-3-nitroaniline, 2-chloro-5-nitro aniline, aniline, 2-chloro-5-nitro, 2-chloro-5-nitrobenzenamine, aniline, 6-chloro-3-nitro, 2-chloro-5-nitrophenylamine, 2-chloro-5-nitro-benzamine, pubchem21904, 3-nitro-6-chloroaniline
PubChem CID 22691
IUPAC Name 2-chloro-5-nitroaniline
SMILES C1=CC(=C(C=C1[N+](=O)[O-])N)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC109575000 SDS View Documents Acros Organics
109575000
500g Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

6283-25-6
172.568
KWIXNFOTNVKIGM-UHFFFAOYSA-N
22691
C1=CC(=C(C=C1[N+](=O)[O-])N)Cl
C6H5ClN2O2
MFCD00007668
benzenamine, 2-chloro-5-nitro, 6-chloro-3-nitroaniline, 2-chloro-5-nitro aniline, aniline, 2-chloro-5-nitro, 2-chloro-5-nitrobenzenamine, aniline, 6-chloro-3-nitro, 2-chloro-5-nitrophenylamine, 2-chloro-5-nitro-benzamine, pubchem21904, 3-nitro-6-chloroaniline
2-chloro-5-nitroaniline

Specifications

Authentic
Yellow
500g
98.5
99%
ClC6H3(NO2)NH2
12, 732
KWIXNFOTNVKIGM-UHFFFAOYSA-N
2-chloro-5-nitroaniline
22691
Powder
2-Chloro-5-nitroaniline, 99%
Glass bottle
118°C to 121°C
6283-25-6
100.0
C6H5ClN2O2
MFCD00007668
benzenamine, 2-chloro-5-nitro, 6-chloro-3-nitroaniline, 2-chloro-5-nitro aniline, aniline, 2-chloro-5-nitro, 2-chloro-5-nitrobenzenamine, aniline, 6-chloro-3-nitro, 2-chloro-5-nitrophenylamine, 2-chloro-5-nitro-benzamine, pubchem21904, 3-nitro-6-chloroaniline
C1=CC(=C(C=C1[N+](=O)[O-])N)Cl
172.568
172.57
≥98.5%
Safety and Handling

Safety and Handling

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Fatal in contact with skin.
Fatal if swallowed.
Fatal if inhaled.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 228-498-7

RTECSNumber : BX1500000

TSCA : TSCA