Learn More
2-Chloro-5-nitroaniline, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1510222
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Chloro-5-nitroaniline | |
117°C to 121°C | |
Weak, Characteristic | |
MFCD00007668 | |
UN2237 | |
benzenamine, 2-chloro-5-nitro, 6-chloro-3-nitroaniline, 2-chloro-5-nitro aniline, aniline, 2-chloro-5-nitro, 2-chloro-5-nitrobenzenamine, aniline, 6-chloro-3-nitro, 2-chloro-5-nitrophenylamine, 2-chloro-5-nitro-benzamine, pubchem21904, 3-nitro-6-chloroaniline | |
C1=CC(=C(C=C1[N+](=O)[O-])N)Cl | |
172.568 | |
172.57 |
6283-25-6 | |
191°C (375°F) | |
C6H5ClN2O2 | |
100 g | |
2208878 | |
KWIXNFOTNVKIGM-UHFFFAOYSA-N | |
2-chloro-5-nitroaniline | |
22691 | |
98% |
Chemical Identifiers
6283-25-6 | |
172.568 | |
KWIXNFOTNVKIGM-UHFFFAOYSA-N | |
22691 | |
C1=CC(=C(C=C1[N+](=O)[O-])N)Cl |
C6H5ClN2O2 | |
MFCD00007668 | |
benzenamine, 2-chloro-5-nitro, 6-chloro-3-nitroaniline, 2-chloro-5-nitro aniline, aniline, 2-chloro-5-nitro, 2-chloro-5-nitrobenzenamine, aniline, 6-chloro-3-nitro, 2-chloro-5-nitrophenylamine, 2-chloro-5-nitro-benzamine, pubchem21904, 3-nitro-6-chloroaniline | |
2-chloro-5-nitroaniline |
Safety and Handling
GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
H300+H310+H330-H373-H500
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLORONITROANILINES
EINECSNumber : 228-498-7
RTECSNumber : BX1500000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only