missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-nitroaniline, 98%

Manufacturer:  Alfa Aesar™ A1510236

 View more versions of this product

Catalog No. AAA1510236



Specifications

Specifications

2-Chloro-5-nitroaniline
Weak, Characteristic
500g
C6H5ClN2O2
UN2237
benzenamine, 2-chloro-5-nitro, 6-chloro-3-nitroaniline, 2-chloro-5-nitro aniline, aniline, 2-chloro-5-nitro, 2-chloro-5-nitrobenzenamine, aniline, 6-chloro-3-nitro, 2-chloro-5-nitrophenylamine, 2-chloro-5-nitro-benzamine, pubchem21904, 3-nitro-6-chloroaniline
C1=CC(=C(C=C1[N+](=O)[O-])N)Cl
172.568
172.57
191°C (375°F)
117°C to 121°C
6283-25-6
MFCD00007668
2208878
KWIXNFOTNVKIGM-UHFFFAOYSA-N
2-chloro-5-nitroaniline
22691
98%

Chemical Identifiers

6283-25-6
172.568
KWIXNFOTNVKIGM-UHFFFAOYSA-N
22691
C1=CC(=C(C=C1[N+](=O)[O-])N)Cl
C6H5ClN2O2
MFCD00007668
benzenamine, 2-chloro-5-nitro, 6-chloro-3-nitroaniline, 2-chloro-5-nitro aniline, aniline, 2-chloro-5-nitro, 2-chloro-5-nitrobenzenamine, aniline, 6-chloro-3-nitro, 2-chloro-5-nitrophenylamine, 2-chloro-5-nitro-benzamine, pubchem21904, 3-nitro-6-chloroaniline
2-chloro-5-nitroaniline
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLORONITROANILINES

RTECSNumber : BX1500000

TSCA : TSCA

Documents