missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-nitro-N-(4-pyridyl)benzamide

$150.22 - $684.70

Chemical Identifiers

CAS 313516-66-4
Molecular Formula C12H8ClN3O3
Molecular Weight (g/mol) 277.664
MDL Number MFCD00121849
InChI Key FRPJSHKMZHWJBE-UHFFFAOYSA-N
Synonym 2-chloro-5-nitro-n-4-pyridinylbenzamide, 2-chloro-5-nitro-n-pyridin-4-yl benzamide, 2-chloro-5-nitro-n-4-pyridyl benzamide, benzamide, 2-chloro-5-nitro-n-4-pyridinyl, 2-chloro-5-nitro-n-4-pyridinyl-benzamide, d03bpg, d08gqc, 2-chloro-5-nitro-n-4-pyridinyl benzamide, benzamide,2-chloro-5-nitro-n-4-pyridinyl, n1-4-pyridyl-2-chloro-5-nitrobenzamide
PubChem CID 2777391
IUPAC Name 2-chloro-5-nitro-N-pyridin-4-ylbenzamide
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(=O)NC2=CC=NC=C2)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ64455MA View Documents Alfa Aesar™
J64455MA
10mg Each for $150.22
 
AAJ64455MF View Documents Alfa Aesar™
J64455MF
50mg Each for $684.70
 
Description

Description

  • A cell-permeable chloro-nitro-benzamido compound that acts as a potent, specific, irreversible, and high-affinity antagonist of PPAR
Specifications

Chemical Identifiers

313516-66-4
277.664
FRPJSHKMZHWJBE-UHFFFAOYSA-N
2777391
C1=CC(=C(C=C1[N+](=O)[O-])C(=O)NC2=CC=NC=C2)Cl
C12H8ClN3O3
MFCD00121849
2-chloro-5-nitro-n-4-pyridinylbenzamide, 2-chloro-5-nitro-n-pyridin-4-yl benzamide, 2-chloro-5-nitro-n-4-pyridyl benzamide, benzamide, 2-chloro-5-nitro-n-4-pyridinyl, 2-chloro-5-nitro-n-4-pyridinyl-benzamide, d03bpg, d08gqc, 2-chloro-5-nitro-n-4-pyridinyl benzamide, benzamide,2-chloro-5-nitro-n-4-pyridinyl, n1-4-pyridyl-2-chloro-5-nitrobenzamide
2-chloro-5-nitro-N-pyridin-4-ylbenzamide

Specifications

Light Sensitive
10mg
C12H8ClN3O3
2-chloro-5-nitro-n-4-pyridinylbenzamide, 2-chloro-5-nitro-n-pyridin-4-yl benzamide, 2-chloro-5-nitro-n-4-pyridyl benzamide, benzamide, 2-chloro-5-nitro-n-4-pyridinyl, 2-chloro-5-nitro-n-4-pyridinyl-benzamide, d03bpg, d08gqc, 2-chloro-5-nitro-n-4-pyridinyl benzamide, benzamide,2-chloro-5-nitro-n-4-pyridinyl, n1-4-pyridyl-2-chloro-5-nitrobenzamide
C1=CC(=C(C=C1[N+](=O)[O-])C(=O)NC2=CC=NC=C2)Cl
277.664
277.66
2-Chloro-5-nitro-N-(4-pyridyl)benzamide
White
313516-66-4
MFCD00121849
FRPJSHKMZHWJBE-UHFFFAOYSA-N
2-chloro-5-nitro-N-pyridin-4-ylbenzamide
2777391
Solid
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C