missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-5-n-propylpyrimidine, 98%

$64.03 - $335.57

Chemical Identifiers

CAS 219555-98-3
Molecular Formula C7H9ClN2
Formula Weight 156.62
MDL Number MFCD00799403
Chemical Name or Material 2-Chloro-5-n-propylpyrimidine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2740903 SDS Alfa Aesar™
H2740903
1g Each for $64.03
AAH2740909 SDS Alfa Aesar™
H2740909
10g Each for $335.57
Specifications Description & Specifications

Chemical Identifiers

219555-98-3
156.62
2-Chloro-5-n-propylpyrimidine
C7H9ClN2
MFCD00799403

Specifications

100°C (3mmHg)
219555-98-3
156.62
2-Chloro-5-n-propylpyrimidine
1g
C7H9ClN2
MFCD00799403
98%

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning