missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Chloro-5-n-pentylpyrimidine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2780809
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Chloro-5-n-pentylpyrimidine | |
1.085 | |
97°C (207°F) | |
1.504 | |
10 g | |
WMARUIMXVALOMF-UHFFFAOYSA-N | |
2-chloro-5-pentylpyrimidine | |
16756526 | |
98% |
154466-62-3 | |
124°C (3 mmHg) | |
C9H13ClN2 | |
MFCD04038093 | |
2-chloro-5-n-pentylpyrimidine, pubchem9038, 5-pentyl-2-chloropyrimidine, 2-chloro-5-pentyl pyrimidine, 2-chloranyl-5-pentyl-pyrimidine | |
CCCCCC1=CN=C(N=C1)Cl | |
184.667 | |
184.67 |
Chemical Identifiers
154466-62-3 | |
184.667 | |
WMARUIMXVALOMF-UHFFFAOYSA-N | |
16756526 | |
CCCCCC1=CN=C(N=C1)Cl |
C9H13ClN2 | |
MFCD04038093 | |
2-chloro-5-n-pentylpyrimidine, pubchem9038, 5-pentyl-2-chloropyrimidine, 2-chloro-5-pentyl pyrimidine, 2-chloranyl-5-pentyl-pyrimidine | |
2-chloro-5-pentylpyrimidine |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only